Automation Pipette Tips for QIAGEN® Workstations, Porex BioProducts
# de catalogue 46300-480
Fournisseur: Thermo Scientific
Spécifications
- Volume:300 µl
- Tip Color:Black
- Packed:96/Rack, 10 Racks
- Sterile:+
- Supplier No.:BLK-180-96RS
Spécifications
A propos de cet article
Compatible with QIAGEN® robotic workstations.
High quality, flexible polypropylene to ensure proper fit for accuracy and precision.
Available in two volume sizes, 300 and 1100 µl. Tips are available with filter and in two different colors, natural and black.
Customers Who Bought This Also Bought