Automation Pipette Tips for Tecan® Workstations, Porex BioProducts
# de catalogue 46300-426
Fournisseur: Thermo Scientific
Spécifications
- Volume:10 µl
- Tip Color:Natural
- Packed:96/Rack, 10 Racks
- Filter:+
- Sterile:+
- Tip Style:With Filter
- Supplier No.:LMF171-96RS-10
Spécifications
A propos de cet article
Tips are compatible with, but not limited to, the Tecan® Genesis workstation.
Volumes range from 10 to 1000 µl. Available in two different colors, natural and black. Also available with or without filter. The flexible polypropylene design provides optimal fit for the workstation. Sterile tips are tray-packaged.
Customers Who Bought This Also Bought