Automation Pipette Tips for Zymark® Tempo™ Workstations, Porex BioProducts
# de catalogue 46300-328
Fournisseur: Thermo Scientific
Spécifications
- Volume:200 µl
- Tip Color:Natural
- Packed:96/Rack, 10 Racks
- Sterile:+
- Supplier No.:160-96RS
Spécifications
A propos de cet article
Tips are compatible with the Zymark automated liquid handling systems.
Designed to ensure reliable operation with RapidPlate® 96, RapidPlate® 96/384, and all Micro Assay modules and workstations. Racked tips have a rigid box design for reliable tip attachment and individual compartments for each tip, preventing cross contamination.
Customers Who Bought This Also Bought