ContactUsLinkComponent
KIMAX*brand Centrifuge Tube
Accessories for High-Strength Round-Bottom Centrifuge Tubes, Glass, Kimble Chase
# de catalogue 46100-830
KIMAX*brand Centrifuge Tube
Accessories for High-Strength Round-Bottom Centrifuge Tubes, Glass, Kimble Chase
# de catalogue 46100-830
Spécifications
  • Description:
    Rubber Adapter Sleeves for 15 mL Tubes
  • Cat. no.:
    46100-830
  • Supplier No.:
    45550-15