ContactUsLinkComponent
Spilfyter* KOLOR-SAFE* Neutralizer
SPILFYTER® KOLORSAFE® 3-Color Change Dry Neutralizer for Base Spills, NPS
# de catalogue 46000-164
Fournisseur:  NPS
Spilfyter* KOLOR-SAFE* Neutralizer
SPILFYTER® KOLORSAFE® 3-Color Change Dry Neutralizer for Base Spills, NPS
# de catalogue 46000-164
Fournisseur:  NPS
Spécifications
  • Description:
    Base Neutralizer
  • Size:
    0.9 kg
  • Cat. no.:
    46000-164
  • Supplier No.:
    450001
Spécifications
A propos de cet article
KOLOR-SAFE® Dry formula provides a precise method of responding to hazardous base spills.
  • Easy application and greater neutralization capacity
  • Concentrated formula
  • Precise method of response
KOLOR-SAFE® Dry formula provides a precise method of responding to hazardous base spills. Concentrated formula allows for easy application and greater neutralization capacity.
Customers Who Bought This Also Bought