SPILFYTER® KOLORSAFE® 3-Color Change Dry Neutralizer for Base Spills, NPS
# de catalogue 46000-164
Fournisseur: NPS
Spécifications
- Description:Base Neutralizer
- Size:0.9 kg
- Cat. no.:46000-164
- Supplier No.:450001
Spécifications
A propos de cet article
KOLOR-SAFE® Dry formula provides a precise method of responding to hazardous base spills.
- Easy application and greater neutralization capacity
- Concentrated formula
- Precise method of response
KOLOR-SAFE® Dry formula provides a precise method of responding to hazardous base spills. Concentrated formula allows for easy application and greater neutralization capacity.
Customers Who Bought This Also Bought