Order Entry
ContactUsLinkComponent
VWR Plastic Bottles
VWR® Large Wide Mouth Sample Bottle, HDPE
# de catalogue 414004-186
Fournisseur:  VWR International
VWR Plastic Bottles
VWR® Large Wide Mouth Sample Bottle, HDPE
# de catalogue 414004-186
Fournisseur:  VWR International

Spécifications

  • Capacity:
    4 L (1 gal.)
  • Cat. no.:
    414004-186
  • Diameter ×H:
    16.3 x 29.8 cm (67/16 x 113/4")

Spécifications

A propos de cet article

Leakproof bottles are ideal for dry chemicals, specimen collecting, and water sampling.

  • Leakproof
  • Easy-to-grip integrated handle
  • HDPE construction

They can be stored in a freezer to −100 °C (−148 °F). The 4 L (1 gal.) bottles feature an integrated handle for better grip, greater safety, and pouring convenience.

Ordering information: Size 100-415 polypropylene screw caps included.
Customers Who Bought This Also Bought