ContactUsLinkComponent
VWR Boston Round Bottles
VWR® Narrow Mouth Laboratory Bottles, Amber HDPE
# de catalogue 414004-168
Fournisseur:  VWR International
VWR Boston Round Bottles
VWR® Narrow Mouth Laboratory Bottles, Amber HDPE
# de catalogue 414004-168
Fournisseur:  VWR International
Spécifications
  • Cap Size:
    38-430
  • Capacity:
    1 L (33.8 oz.)
  • Cat. no.:
    414004-168
Spécifications
A propos de cet article
Bottles are for general laboratory use and for storing, shipping, and packing liquids, especially light-sensitive materials or photosensitive samples.
  • Amber coloring protects light-sensitive contents
  • High-density polyethylene fabrication
  • Excellent chemical resistance
Can be stored in freezer to −100 °C (−148 °F), and have excellent chemical resistance to most acids, bases, and alcohols. Amber polypropylene screw closure included. Bottles comply with current USP requirements for maximum light transmission.
Customers Who Bought This Also Bought