VWR®, Graduated Beakers, Polypropylene
Catalog # 414004-148
Supplier: VWR International
Specifications
- Graduation range:10 - 250 ml
- Capacity:250 ml (8.5 oz.)
- Cat. no.:414004-148
Specifications
About this item
Beakers feature a pouring spout for quick and easy dispensing.
- Withstand temperatures up to 121°C
- Shatter-resistant
- Easy-to-read graduations
- Transparent for clear content visibility
Highly resistant to strong acids. Stackable. Steam autoclavable.
Customers Who Bought This Also Bought