VWR®, Graduated Beakers, Polypropylene
# de catalogue 414004-147
Fournisseur: VWR International
Spécifications
- Capacity:150 ml (5 oz.)
- Graduation range:5 - 150 ml
Spécifications
A propos de cet article
Beakers feature a pouring spout for quick and easy dispensing.
- Withstand temperatures up to 121°C
- Shatter-resistant
- Easy-to-read graduations
- Transparent for clear content visibility
Highly resistant to strong acids. Stackable. Steam autoclavable.
Customers Who Bought This Also Bought