VWR® Disposable Transfer Pipettes
Catalog # 414004-042
Supplier: VWR International
Specifications
- Sterility:Sterile
- Capacity:5.0 ml
- Description:Extended fine tip, large bulb
- Packed:20/bag
- Overall Length:15.2 cm (6")
- Bulb Draw:3.3 ml
- Drops per mL:50
Specifications
About this item
These unbreakable all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials.
- Will not shatter or break
- Can be used in liquid nitrogen
- Nontoxic and inert
- No bulb to insert or remove
- Uniform drop size
Molded from see-through low density polyethylene, they are inert to biological fluids and most acids. The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Transfer pipettes can be sealed and refrigerated, and will work well whenever there is a need for quick, safe fluid transfer. Can be gas sterilized.
Frequently Bought Together