VWR® Disposable Transfer Pipettes, Graduated
# de catalogue 414004-035
Fournisseur: VWR International
Spécifications
- Capacity:5.8 ml
- Sterility:Non sterile
- Description:Large Bulb
- Overall Length:15.5 cm (6⅛")
- Graduations:0.25 mL
- Bulb Draw:3.4 ml
- Drops per mL:25
- Maximum Graduation Volume:1 ml
Spécifications
A propos de cet article
VWR® Disposable Transfer Pipettes are ideal for quick and safe fluid transfer.
- Will not shatter or break
- Can be used in liquid nitrogen
- Nontoxic and inert
- No bulb to insert or remove
- Uniform drop size
These unbreakable all-in-one graduated transfer pipettes eliminate the hazard of broken glass and exposure to infectious materials. Molded from see-through low density polyethylene, pipettes are inert to biological fluids and most acids, and can be sealed and refrigerated. The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Ideal for quick, safe transfer of fluids. Transfer pipettes can be gas sterilized. Manufactured from LDPE, they can be sealed, refrigerated, and gas sterilized.
Customers Who Bought This Also Bought