Orion™ ISE Calibration Standards, Thermo Scientific
# de catalogue 34184-174
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Concentration:100 ppm
- Description:Residual Chlorine Standard
- Size:475 ml
- Cat. no.:34184-174
- Supplier No.:977007
Spécifications
A propos de cet article
Take reproducible ISE measurements with Orion™ ISE Calibration Standards
For use with ion selective electrodes from ammonia to water hardness.
Ordering information: The Fluoride standard bulk pack includes 4x475mL each of 1ppm fluoride standard premixed with TISAB II and 10ppm fluoride standard premixed with TISAB II.
Packaging: All standards are supplied in 1 pint (475 mL) bottles, unless otherwise noted
Customers Who Bought This Also Bought