Order Entry
ContactUsLinkComponent
Value Ear Muffs
ValueMuff™ Earmuffs, Elvex®
# de catalogue 33200-000
Fournisseur:  Elvex
Small Business Enterprise
Value Ear Muffs
ValueMuff™ Earmuffs, Elvex®
# de catalogue 33200-000
Fournisseur:  Elvex
Small Business Enterprise

Spécifications

  • NRR:
    25 dB
  • Description:
    Earmuffs ValueMuff™
  • Cat. no.:
    33200-000
  • Supplier No.:
    HB-25

Spécifications

A propos de cet article

Dielectric, comfortable, lightweight muff at an economical price

  • Soft, pliable Superseal™ cushions and low headband force of 2.4lbs
  • Flexible slide clips for easy adjustment
  • Good attenuation
Certifications: Meets ANSI 53.19-1974 standard.
Customers Who Bought This Also Bought