SP Bel-Art Rotary Separatory Funnel Rack, Bel-Art Products, a part of SP
# de catalogue 30534-900
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Ø:35 cm (13 3/4")
- Description:Rotary Separatory Funnel Rack
- Cat. no.:30534-900
- Supplier No.:189640000
Spécifications
A propos de cet article
Rack consists of a rectangular base, screw-in column, and a support disc held in place by a screw clamp.
- Fully adjustable support disc
- Open front for easy removal and placement of separatory funnels
- Insert clips accommodate various sizes of funnels
The support disc is fully adjustable for different heights up to 53.3 cm (21"). Designed to accommodate a maximum of six 500 mL or 1000 mL funnels.
Delivery: Funnel rack includes 18 insert clips used to accommodate different sized separatory funnels. Shipped unassembled.
Customers Who Bought This Also Bought