Flexible Funnels, Hutzler Manufacturing
# de catalogue 30251-021
Fournisseur: Hutzler Manufacturing
Spécifications
- Material:PP
- Top I.D.:68 mm
- Stem O.D.:9.7 mm
- Stem Length:20.2 mm
- Supplier No.:802-PP
Spécifications
A propos de cet article
Thin, flexible
Ribbed to permit air to escape as fluid flows in. Color: Translucent white. 30251-101 has wide neck for larger diameter containers.
Customers Who Bought This Also Bought