Order Entry
ContactUsLinkComponent
VWR* Media Bottle, Polycarbonate, Graduated
VWR® Boston Round Media Bottles, PC
# de catalogue 30180-030
Fournisseur:  VWR International
VWR* Media Bottle, Polycarbonate, Graduated
VWR® Boston Round Media Bottles, PC
# de catalogue 30180-030
Fournisseur:  VWR International

Spécifications

  • Cap Size:
    38-430
  • Capacity:
    500 ml (16.9 oz.)
  • Graduation Interval:
    50 ml
  • Graduation range:
    50–500 mL
  • Cat. no.:
    30180-030

Spécifications

A propos de cet article

VWR® polycarbonate resin is an amorphous engineering thermoplastic, characterized by outstanding mechanical, optical, electrical, and thermal properties.

  • Polycarbonate resin construction
  • Autoclavable
  • Optimal for preparation of terminally sterile fluids

The Lexan® resins employed are environmentally conforming to flame retardancy, scratch resistance, toughness, heat resistance, weatherability, biocompatibility, optical quality, and compliance with stringent FDA and USP requirements.

All bottles are manufactured and packaged in a Class 10K cleanroom in an ISO 9001 compliant facility, and do not require rinsing prior to first use.

A safe alternative to glass vessels, these media bottles are ideal for steam sterilization of all biologicals, liquids, reagents, media, buffers, and aqueous solutions. Media bottles withstand repeated autoclaving, and are optimal for all microbial culture media production and preparation of terminally sterile fluids.

Ordering information: Bottles are supplied nonsterile with standard blue autoclavable caps attached. Packaged in multi-barrier elastomer packaging. Caps are available with custom color options on bulk orders. For additional cap selections, see 82013-158 series.
Customers Who Bought This Also Bought