Nalgene® Polypropylene Locking Forceps, Thermo Scientific
Catalog # 30049-122
Supplier: Thermo Fisher Scientific
Specifications
- Description:Plastic Forceps
- Cat. no.:30049-122
- Supplier No.:6320-0010
Specifications
About this item
All-plastic forceps with double-action hinges for extra leverage and long life
Scissors-type handles with ratchet for locking tips shut. Interlocking teeth for tight grip. Provide a useful clamp for chromatography columns. Durable blue polypropylene is autoclavable for repeated use. Ideal for economical, disposable applications.
Customers Who Bought This Also Bought