ContactUsLinkComponent
NALGENE* Locking Forceps
Nalgene® Polypropylene Locking Forceps, Thermo Scientific
Catalog # 30049-122
NALGENE* Locking Forceps
Nalgene® Polypropylene Locking Forceps, Thermo Scientific
Catalog # 30049-122
Specifications
  • Description:
    Plastic Forceps
  • Cat. no.:
    30049-122
  • Supplier No.:
    6320-0010
Specifications
About this item
All-plastic forceps with double-action hinges for extra leverage and long life
Scissors-type handles with ratchet for locking tips shut. Interlocking teeth for tight grip. Provide a useful clamp for chromatography columns. Durable blue polypropylene is autoclavable for repeated use. Ideal for economical, disposable applications.
Customers Who Bought This Also Bought