Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
# de catalogue 29417-048
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:1 L
- Cat. no.:29417-048
- Rubber Stopper Size:8
- Supplier No.:DS4101-1000
Spécifications
A propos de cet article
One-piece molded for strength and stability
Withstand full vacuum to 762mm Hg (30in. Hg). Tubulation at 45° angle prevents tipping. Oval marking area. Barbed adapter accepts 6.4mm (1/4") to 9.5mm (3/8") I.D. tubing. Autoclavable.
Customers Who Bought This Also Bought