Order Entry
ContactUsLinkComponent
PYREX* brand Erlenmeyer Flasks, Graduated, Wide Mouth, Heavy-Duty Rim
PYREX® Erlenmeyer flasks, Wide Neck, Heavy-Duty Rim, Corning
Catalog # 29140-089
Supplier:  Corning
Product available on GSA Advantage®
undefined
PYREX® Erlenmeyer flasks, Wide Neck, Heavy-Duty Rim, Corning
Catalog # 29140-089
Supplier:  Corning
Product available on GSA Advantage®

Specifications

  • Graduation Interval:
    50 ml
  • Graduation Range:
    250 - 1000 ml
  • Capacity:
    1000 ml
  • Description:
    Pyrex® wide-mouth erlenmeyer flask
  • Cat. no.:
    29140-089
  • Stopper No.:
    11
  • Supplier No.:
    5100-1L

Specifications

About this item

Heavy-duty rim flasks are especially recommended for use as titration flasks.

  • White enamel graduation to show the approximate capacity
  • With extra large marking spot
  • Good thermal and chemical resistance

Designed for long life and maximum safety, these flasks have a wide mouth, as indicated by stopper openings.

Frequently Bought Together