Nalgene® Erlenmeyer Flasks, Polycarbonate, Thermo Scientific
# de catalogue 29136-944
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:250 ml
- Cat. no.:29136-944
- Stopper No.:6
- Supplier No.:4103-0250
Spécifications
A propos de cet article
Graduated flasks feature excellent clarity, toughness, and strength.
Molded-in graduations. Autoclavable, but repeated autoclaving may reduce mechanical strength.
Customers Who Bought This Also Bought