ContactUsLinkComponent
NALGENE* Erlenmeyer Flask, Polycarbonate
Nalgene® Erlenmeyer Flasks, Polycarbonate, Thermo Scientific
# de catalogue 29136-944
NALGENE* Erlenmeyer Flask, Polycarbonate
Nalgene® Erlenmeyer Flasks, Polycarbonate, Thermo Scientific
# de catalogue 29136-944
Spécifications
  • Capacity:
    250 ml
  • Cat. no.:
    29136-944
  • Stopper No.:
    6
  • Supplier No.:
    4103-0250
Spécifications
A propos de cet article
Graduated flasks feature excellent clarity, toughness, and strength.
Molded-in graduations. Autoclavable, but repeated autoclaving may reduce mechanical strength.
Customers Who Bought This Also Bought