Accessories for Nalgene® Rapid-Flow™ Bottle-Top Vacuum Filters, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
# de catalogue 28199-402
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:Glass Prefilter for 90 mm Diameter Membranes. Thin, Nonsterile, Very Low Levels of Extractables, For Pre-Filtering Viscous or Heavily Particulated Fluids
- Cat. no.:28199-402
- Supplier No.:DS0281-9000