ContactUsLinkComponent
Glass Prefilter
Accessories for Nalgene® Rapid-Flow™ Bottle-Top Vacuum Filters, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
# de catalogue 28199-402
Accessories for Nalgene® Rapid-Flow™ Bottle-Top Vacuum Filters, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
# de catalogue 28199-402
Spécifications
  • Description:
    Glass Prefilter for 90 mm Diameter Membranes. Thin, Nonsterile, Very Low Levels of Extractables, For Pre-Filtering Viscous or Heavily Particulated Fluids
  • Cat. no.:
    28199-402
  • Supplier No.:
    DS0281-9000