ContactUsLinkComponent
Reusable Bottle-Top Filter, PSF (Fits 45 mm neck), non sterile
Nalgene® Polysulfone Reusable Bottle Top Filters, Thermo Scientific
# de catalogue 28199-325
Reusable Bottle-Top Filter, PSF (Fits 45 mm neck), non sterile
Nalgene® Polysulfone Reusable Bottle Top Filters, Thermo Scientific
# de catalogue 28199-325
Spécifications
  • Capacity:
    500 ml (upper chamber)
  • Description:
    Reusable bottle top filter
  • Cat. no.:
    28199-325
  • Neck Diameter:
    45
  • Filtration Area:
    13.3 cm4
  • Supplier No.:
    DS0320-5045
Spécifications
A propos de cet article
Nalgene® Polysulfone Reusable Bottle Top Filters offer the convenience of a bottle top filter with the cost savings and waste reduction of a reusable filter housing.
The filters feature a removable sterilization membrane support plate designed to provide maximum flow rate and throughput, as well as a vacuum port on the lower half of the assembly and molded-in graduations. The polypropylene tubing adapter can be stuffed with cotton for sterile venting during autoclaving.
Nalgene® Polysulfone Reusable Bottle Top Filters offer the convenience of a bottle top filter with the cost savings and waste reduction of a reusable filter housing.
The filters feature a removable sterilization membrane support plate designed to provide maximum flow rate and throughput, as well as a vacuum port on the lower half of the assembly and molded-in graduations. The polypropylene tubing adapter can be stuffed with cotton for sterile venting during autoclaving.
The deep threads screw securely onto the bottle for convenient filling and vacuum filtration.
Customers Who Bought This Also Bought