Spécifications
- Capacity:500 ml (upper chamber)
- Description:Reusable bottle top filter
- Cat. no.:28199-325
- Neck Diameter:45
- Filtration Area:13.3 cm4
- Supplier No.:DS0320-5045
Spécifications
A propos de cet article
Nalgene® Polysulfone Reusable Bottle Top Filters offer the convenience of a bottle top filter with the cost savings and waste reduction of a reusable filter housing.
The filters feature a removable sterilization membrane support plate designed to provide maximum flow rate and throughput, as well as a vacuum port on the lower half of the assembly and molded-in graduations. The polypropylene tubing adapter can be stuffed with cotton for sterile venting during autoclaving.
Nalgene® Polysulfone Reusable Bottle Top Filters offer the convenience of a bottle top filter with the cost savings and waste reduction of a reusable filter housing.
The filters feature a removable sterilization membrane support plate designed to provide maximum flow rate and throughput, as well as a vacuum port on the lower half of the assembly and molded-in graduations. The polypropylene tubing adapter can be stuffed with cotton for sterile venting during autoclaving.
The deep threads screw securely onto the bottle for convenient filling and vacuum filtration.