Nalgene® Laboratory Notebooks, Thermo Scientific
# de catalogue 28196-344
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cover Color:Burgundy
- Ruling:Ruled, 3 lines/in.
- Cat. no.:28196-344
- Length:281 mm (11")
- Width:216 mm (8.5")
- Supplier No.:6301-2000
Spécifications
A propos de cet article
Notebooks feature sewn-in paper pages with either horizontal ruled lines or grid
Pages have spaces for dates, signatures of workers and witnesses, and other necessary information for securing patent protection. Polyethylene cover is waterproof. Notebooks have 100 numbered pages.
Customers Who Bought This Also Bought