Order Entry
ContactUsLinkComponent
Nalgene 50mm Inline Syringe Filter
Nalgene® Syringe/In-Line Filters, PTFE, 50 mm, Thermo Scientific
# de catalogue 28160-810
Nalgene 50mm Inline Syringe Filter
Nalgene® Syringe/In-Line Filters, PTFE, 50 mm, Thermo Scientific
# de catalogue 28160-810

Spécifications

  • Pore size:
    0.2 µm
  • Cat. no.:
    28160-810
  • Supplier No.:
    DS0222-0020

Spécifications

A propos de cet article

For filtration of air, gases, and aggressive chemicals, including acids and non-aqueous solvents.

Helps meet OSHA standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens. Useful for benchtop fermentors, in-line gas filtration for sterile air or CO₂, and protection of vacuum pumps from damage by aqueous solution. Diameter: 50 mm. Filter area: 20 cm². Sample volume size from 200 ml to 5 L. Polypropylene housing. Stepped hose inlet/outlet barbs accept 6 to 9mm (¼ to ¾") int.Ø. tubing. Nonsterile. Autoclavable.

Customers Who Bought This Also Bought