Coplin Staining Jar with Cover, Wheaton
# de catalogue 25460-000
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Small Business Enterprise
Spécifications
- Dimensions:3 I.D. x 7.6 H cm (13/16 x 3")
- Supplier No.:900470
Spécifications
A propos de cet article
Heavy soda lime glass, with ground-on cover and wide, flat base, for convenient manipulation of sections attached to slides
Holds ten 75x25mm (3x1") slides.
Customers Who Bought This Also Bought