VWR® Disposable Aluminum Smooth-Wall Weighing Dishes
Catalog # 25433-089
Supplier: VWR International
Specifications
- Capacity:80 ml
- I.D.×D:7×1.6 cm (2¾×⅝")
Specifications
About this item
These smooth-wall, flat-bottom dishes are ideal for general-purpose weighing, dispensing, storing, sampling, and testing applications.
- Ideal for general-purpose applications
- Contoured design
- Disposable
- Aluminum construction
Contoured to facilitate stacking, weighing, and dispensing.
Frequently Bought Together