ContactUsLinkComponent
Vwr* Tri-Pour* Graduated Disposable Beakers
VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
# de catalogue 25384-156
Fournisseur:  VWR International
Vwr* Tri-Pour* Graduated Disposable Beakers
VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
# de catalogue 25384-156
Fournisseur:  VWR International
Spécifications
  • Capacity:
    400 ml (14 oz.)
Spécifications
A propos de cet article
Transparent polypropylene beaker with three pouring lips for dripless dispensing.
  • Polypropylene construction
  • Transparent
  • Graduated markings
  • Disposable design
Graduated, with accuracy of ±5%. May also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents. Write-on cardboard caps ensure spillproof, leak-resistant closure.
Customers Who Bought This Also Bought