Order Entry
Canada
ContactUsLinkComponent
Spectra/Por® 1 Dialysis Membranes, MWCO 6000 to 8000, Spectrum® Laboratories
Catalogue # 25218-038
Fournisseur:  Spectrum Laboratories
Small Business Enterprise
undefined
Spectra/Por® 1 Dialysis Membranes, MWCO 6000 to 8000, Spectrum® Laboratories
Catalogue # 25218-038
Fournisseur:  Spectrum Laboratories
Supplier Number:  132650
Small Business Enterprise

Spécifications

  • Ø:
    14.6 mm
  • Length:
    30.5 m
  • Cat. no.:
    25218-038
  • Flat Width:
    23 mm
  • Supplier No.:
    132650

Spécifications

À propos de cet article

Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, therefore these membranes are perfectly suited for a broad range of laboratory dialysis.

The clear and flexible membrane type with a symmetric porosity is economical and sturdy, ideal for desalting, buffer exchanges, macromolecular purifications and other general applications. Possessing a broad range of chemical compatibilities, Standard RC membranes can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols, and some mild or dilute organic solvents, including DMSO.

Frequently Bought Together