ContactUsLinkComponent
UPRIGHT CLEANING BRUSH SS
Stainless Steel Bristle and Wood Handle Applicator Brush, Gordon Brush
# de catalogue 22940-758
Fournisseur:  Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Stainless Steel Bristle and Wood Handle Applicator Brush, Gordon Brush
# de catalogue 22940-758
Fournisseur:  Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Bristle Material:
    Stainless steel
  • Brush Head Diameter:
    0.006"
  • Handle Material:
    Wood
  • Description:
    Applicator Brush with Stainless Steel Bristle and Wood Handle
  • Brush Head Width:
    5/16"
  • Cat. no.:
    22940-758
  • No. of Rows:
    2×6
  • Handle Length:
    415/16"
  • Width:
    11/16"
  • Supplier No.:
    WA12SSG-12
  • Brush Head Length:
    7/8"
  • Trim Length:
    ½"
Spécifications
A propos de cet article
A heavy-duty scrubbing duster brush that works well for getting into tight areas.
  • Known as a typewriter brush
Not recommended for use on circuit boards or SMTs as the stainless steel wire may cause unwanted scratching.
Customers Who Bought This Also Bought