Specifications
- Description:Carbon Steel Blade
- Cat. no.:21909-612
- Blade Size:#11
- Supplier No.:4-111
Specifications
About this item
Carbon steel and stainless steel surgical blades feature razor-sharp cutting edges for superior cutting and clean incisions
Blades are for use with No. 3 or No. 4 scalpel handles. No. 3 scalpel handles accommodate blade numbers 10 through 15C. No. 4 scalpel handles accommodate blade numbers 20 through 25. Size is clearly marked on each blade. Individually packaged in sealed, peel-apart pouches. Five-year shelf life.