Spécifications
- Length:16.5 cm (6 1/2")
- Cat. no.:21909-176
- Supplier No.:8-16TC
Spécifications
A propos de cet article
Needle Holders with Suture Scissors
A close examination of Miltex Needle Holders, either Standard or Tungsten Carbide, will reveal meticulously crafted surgical tools that will perform precisely as intended. Smoothly beveled box locks and jaw edges protect delicate suture material from snags or cutting. The jaw surface is milled to ensure a secure grasp and to prevent needle slippage or rotation.