ContactUsLinkComponent
CRB-N-SRT OLSEN HEGAR NH 6.5
Olsen-Hegar Needle Holders, Integra™ Miltex®
# de catalogue 21909-176
Fournisseur:  Miltex
CRB-N-SRT OLSEN HEGAR NH 6.5
Olsen-Hegar Needle Holders, Integra™ Miltex®
# de catalogue 21909-176
Fournisseur:  Miltex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Length:
    16.5 cm (6 1/2")
  • Cat. no.:
    21909-176
  • Supplier No.:
    8-16TC
Spécifications
A propos de cet article
Needle Holders with Suture Scissors
A close examination of Miltex Needle Holders, either Standard or Tungsten Carbide, will reveal meticulously crafted surgical tools that will perform precisely as intended. Smoothly beveled box locks and jaw edges protect delicate suture material from snags or cutting. The jaw surface is milled to ensure a secure grasp and to prevent needle slippage or rotation.
Customers Who Bought This Also Bought