Specifications
- Size:4×3.8 L (1 gal.)
- Cat. no.:21839-066
- Packaging:Bottle
- pH:11
- Supplier No.:1701
Specifications
About this item
Detergent 8® is a nonionic, detergent for labware washers, PC board washers, power-spray systems, ultrasonic and manual cleaning. Removes oils, dirt, grime, residues, fluxes, chemicals, resins rosins, and fluxes from PC circuit boards, electronic parts, phosphate sensitive labware and nuclear contaminated equipment with no interfering conductive residues. Excellent replacement for solvent cleaning.
- Replace solvent cleaning
- Concentrated, biodegradable and readily disposable
- Penetrating wetting power to save you time
- Free rinsing to give you reliable results and no interfering residues
- Contains no phosphate, sulfur, chlorine, metal cation, halide, borate, silicate, carbonate, chlorocarbon, fluorocarbon or chelating ingredients
- Dilute 3:100
Cleaning method: Soak, brush, sponge, cloth, ultrasonic, ware washer, spray washer, pressure washer and flow through CIP (clean-in-place).
Corrosion inhibited formulation recommended for glass, metal, stainless steel, ceramic, plastic, and circuit boards. Can be used, with prompt rinsing, on soft metals such as copper, aluminum, silver, and gold. Aluminum can darken and mixed metal galvanic reactions can occur.
Note: Corrosion testing is advisable.