ContactUsLinkComponent
Detergent 8* Phosphate-Free Liquid Detergent
Detergent 8® Low-Foaming Ion-Free Detergents
Catalog # 21839-066
Supplier:  Alconox
Detergent 8* Phosphate-Free Liquid Detergent
Detergent 8® Low-Foaming Ion-Free Detergents
Catalog # 21839-066
Supplier:  Alconox
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    4×3.8 L (1 gal.)
  • Cat. no.:
    21839-066
  • Packaging:
    Bottle
  • pH:
    11
  • Supplier No.:
    1701
Specifications
About this item
Detergent 8® is a nonionic, detergent for labware washers, PC board washers, power-spray systems, ultrasonic and manual cleaning. Removes oils, dirt, grime, residues, fluxes, chemicals, resins rosins, and fluxes from PC circuit boards, electronic parts, phosphate sensitive labware and nuclear contaminated equipment with no interfering conductive residues. Excellent replacement for solvent cleaning.
  • Replace solvent cleaning
  • Concentrated, biodegradable and readily disposable
  • Penetrating wetting power to save you time
  • Free rinsing to give you reliable results and no interfering residues
  • Contains no phosphate, sulfur, chlorine, metal cation, halide, borate, silicate, carbonate, chlorocarbon, fluorocarbon or chelating ingredients
  • Dilute 3:100
Cleaning method: Soak, brush, sponge, cloth, ultrasonic, ware washer, spray washer, pressure washer and flow through CIP (clean-in-place).
Corrosion inhibited formulation recommended for glass, metal, stainless steel, ceramic, plastic, and circuit boards. Can be used, with prompt rinsing, on soft metals such as copper, aluminum, silver, and gold. Aluminum can darken and mixed metal galvanic reactions can occur.
Note: Corrosion testing is advisable.
Certifications: FDA certified.
Customers Who Bought This Also Bought