ContactUsLinkComponent
Liquinox* Phosphate-Free Liquid Detergent
Liquinox® Critical Cleaning Liquid Detergents
# de catalogue 21837-027
Fournisseur:  Alconox
Liquinox* Phosphate-Free Liquid Detergent
Liquinox® Critical Cleaning Liquid Detergents
# de catalogue 21837-027
Fournisseur:  Alconox
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    4×3.8 L (1 gal.)
  • pH:
    8.5
  • Supplier No.:
    1201
  • Packing Type:
    Bottle
Spécifications
A propos de cet article
Liquinox is a concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water.
  • Replaces corrosive acids and hazardous solvents
  • Phosphate free, biodegradable and readily disposable
  • Free rinsing to give you reliable results and no interfering residues
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 1:100
It can be used to clean Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, wine glasses, clean rooms, medical devices, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, pipes, tanks and reactors. Passes inhibitory residue test for water analysis.
And it can be used to remove soil, grit, grime, slime, grease, oils, blood, tissue, particulates, deposits, chemical and solvents.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, cement and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Used for art restoration. Corrosion testing may be advisable.
Cleaning method: Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace.
Certifications: USDA authorized. FDA certified.
Caution: Will foam - not for spray or machine use.
Customers Who Bought This Also Bought