ContactUsLinkComponent
DET-O-JET DETERGENT 1GL
Detojet® Low Foaming Critical Detergents
# de catalogue 21835-258
Fournisseur:  Alconox
DET-O-JET DETERGENT 1GL
Detojet® Low Foaming Critical Detergents
# de catalogue 21835-258
Fournisseur:  Alconox
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    4×3.8 L (1 gal.)
  • Packaging:
    Bottle
  • pH:
    12.4
  • Supplier No.:
    1601
Spécifications
A propos de cet article
Detojet® is a low foaming, heavy duty alkaline detergent for use in labware washers, parts washers, power-spray systems, and ultrasonic cleaning systems. Concentrated, highly emulsifying and penetrating formula is free rinsing, and leaves no interfering residues. Protects interior working parts of washers. Completely soluble in hot or cold, hard or soft water. Protects and lubricates working parts of washers.
  • Biodegradable and disposable
  • Dilute 1:100
  • High performance results from cleaning machinery
For removing soil, grit, organic contaminants, inorganic residue, chemicals, and solvents from glass, stainless steel, ceramic, cement, and alkali-resistant plastics and metals. Ideal for mechanical washers, power-spray systems, and ultrasonic cleaners, including those equipped with automatic feed and proportioning devices.
They can be used to clean hospital instruments, laboratory ware, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors.
Cleaning method: Laboratory warewashers, dishwashers, washer-sterilizers, washersanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners flow through CIP (clean-in-place) and manual cleaning if proper skin and eye protection is used.
Recommended for glass, stainless steel, ceramic, cement and alkali resistant plastics and metals such as stainless steel.
Note: Corrosion testing is advisable.
Certifications: Authorized by USDA for use in federally inspected meat and poultry plants. FDA certified.
Customers Who Bought This Also Bought