Spécifications
- Size:4×3.8 L (1 gal.)
- Packaging:Bottle
- pH:12.4
- Supplier No.:1601
Spécifications
A propos de cet article
Detojet® is a low foaming, heavy duty alkaline detergent for use in labware washers, parts washers, power-spray systems, and ultrasonic cleaning systems. Concentrated, highly emulsifying and penetrating formula is free rinsing, and leaves no interfering residues. Protects interior working parts of washers. Completely soluble in hot or cold, hard or soft water. Protects and lubricates working parts of washers.
- Biodegradable and disposable
- Dilute 1:100
- High performance results from cleaning machinery
For removing soil, grit, organic contaminants, inorganic residue, chemicals, and solvents from glass, stainless steel, ceramic, cement, and alkali-resistant plastics and metals. Ideal for mechanical washers, power-spray systems, and ultrasonic cleaners, including those equipped with automatic feed and proportioning devices.
They can be used to clean hospital instruments, laboratory ware, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors.
Cleaning method: Laboratory warewashers, dishwashers, washer-sterilizers, washersanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners flow through CIP (clean-in-place) and manual cleaning if proper skin and eye protection is used.
Recommended for glass, stainless steel, ceramic, cement and alkali resistant plastics and metals such as stainless steel.
Note: Corrosion testing is advisable.