ContactUsLinkComponent
Alcotabs* Tablet Detergent
Alcotabs® Critical Cleaning Detergent Tablets
# de catalogue 21834-109
Fournisseur:  Alconox
Alcotabs* Tablet Detergent
Alcotabs® Critical Cleaning Detergent Tablets
# de catalogue 21834-109
Fournisseur:  Alconox
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    6×100 tablet
  • Packaging:
    Bottle
  • pH:
    6.5
  • Supplier No.:
    1500
Spécifications
A propos de cet article
Effervescent detergent tablet for lab pipette cleaning. Maximum cleaning in syphon pipette washers to clean pipettes and tubes. Slow release bubble action and free rinsing produce reliable results and no interfering residues.
  • Concentrated
  • Biodegradable and readily disposable
  • Free rinsing to give you reliable results and no interfering residues
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
Special slow release tablet formulation for pipette cleaning. Maximum cleaning performance from your syphon pipette washers.
It can be used to clean pipettes and tubes with syphon pipette washers in hospitals, clinical, educational, R&D and industrial laboratories and cosmetics manufacturing.
And it can be used to remove soil, grit, grime, grease, fats, oils, blood, tissue, salt deposits, particulates, chemicals, solvents and radioisotopes.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Corrosion testing may be advisable.
Cleaning method: Syphon pipette and tube rinser.
Certifications: USDA authorized. FDA certified.
Packaging: Supplied in a bottle of 100 tablets.
Customers Who Bought This Also Bought