VWR® Talon® Jumbo Holders
# de catalogue 21572-556
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:Clamp Holder
- Material:Aluminum
- Cat. no.:21572-556
Spécifications
A propos de cet article
Holder secures clamps to lab frames and ring stands and accepts rods up to 21mm (13/16") in diameter.
- Electropolished stainless steel or aluminum construction
- Secure support of clamps
- Accepts rods of a range of diameters
Jumbo holders provide sturdy attachments of clamps to lab frames and ring stands.
Customers Who Bought This Also Bought