ContactUsLinkComponent
PCR Tube Rack
96-Well Preparation Rack with Cover, Bio Plas
# de catalogue 21150-224
Fournisseur:  Bio Plas
PCR Tube Rack
96-Well Preparation Rack with Cover, Bio Plas
# de catalogue 21150-224
Fournisseur:  Bio Plas
Spécifications
  • Color:
    Yellow
  • Description:
    96-Well Preparation Rack
  • Cat. no.:
    21150-224
  • Supplier No.:
    0034F
Spécifications
A propos de cet article
High quality
  • Designed for preparation and storage
  • Racks for use with a variety of microcentrifuge tubes
Manufactured of polypropylene, these 96-well (8x12 format) racks are suitable for autoclaving. These one sided racks are suitable for 0.2 ml PCR tubes with strip caps. The snap-on cover stays in place during transportation and storage. Racks are able to be securely stacked.
Made in the USA.
Customers Who Bought This Also Bought