VWR® Centrifuge Tubes, Polystyrene
Catalogue # 21008-197
Fournisseur: VWR International
Spécifications
- Cap Type:Flat cap
- Sterility:Sterile
- Volume:15 ml
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001/Certificate No.: 04923E00243R0M
- Cat. no.:21008-197
- Packaging:125/bag, 4 bags/case
- Maximum RCF:3000×g
Spécifications
À propos de cet article
These tubes are made of optically clear PS and feature distinct molded graduations.
- Colorless and transparent are perfect for biological reaction experiments
- RNase-, DNase-, and Endotoxin-Free
- Leak-proof
- Not autoclavable
- Sterile tubes are sterilized by irradiation
- During the freezing process, it is better to use plastic rack
- During high-speed centrifugation, it is better to use suitable rotor adapters
The flat surface of the polypropylene cap provides an at-a-glance identification area.
Certifications: Manufactured in a class 100000 room environment. Manufactured under ISO13485:2016 and ISO9001:2015 quality management. Meet the requirements of RoHS.
Ordering information: Choose from tubes and caps supplied in Earth Friendly cardboard racks, bag packed, or bulk packed.
Frequently Bought Together