Order Entry
ContactUsLinkComponent
NO-WIRE, GRIP RACK, PP, 40 PLACES
SP Bel-Art No-Wire Submersible Plastic Test Tube Grip Racks, Bel-Art Products, a part of SP
# de catalogue 20907-942
SP Bel-Art No-Wire Submersible Plastic Test Tube Grip Racks, Bel-Art Products, a part of SP
# de catalogue 20907-942

Spécifications

  • No. of Holes:
    40 (4×10)
  • Cat. no.:
    20907-942
  • W×D×H:
    24.8×10.5×7 cm (9³/₄×4¹/₈×2³/₄")
  • For Tube O.D.:
    16 - 20 mm
  • Supplier No.:
    187490002

Spécifications

A propos de cet article

Retainer fingers hold tubes firmly in the rack plates with no fear of dislodging, making them perfect for both decanting and water bath work.

  • Fingers allow simultaneous decanting to save valuable time
  • Specially formulized plastic allows the rack to submerge in water baths
  • Fingers secure tubes while in water baths to eliminate the risk of diluting or spilling
  • Features end plates for labeling as well as numbered and lettered rows for tube identification
  • Fold up sides lock firmly into the grip decks; assembly takes only seconds
  • Unloaded racks are stackable

Tube size indicated is for grip function, smaller tubes may fit for storage purposes.

Customers Who Bought This Also Bought