Order Entry
Canada
ContactUsLinkComponent
Thin-Wall Micro Reaction Tubes and Strip Caps, 0.2 ml, Bio Plas
Catalogue # 20170-837
Fournisseur:  Bio Plas
undefined
Thin-Wall Micro Reaction Tubes and Strip Caps, 0.2 ml, Bio Plas
Catalogue # 20170-837
Fournisseur:  Bio Plas
Supplier Number:  5030-1

Spécifications

  • Volume:
  • Color:
    Natural
  • Description:
    8 Strip Caps for 0.2mL Micro Tubes
  • Cat. no.:
    20170-837
  • Supplier No.:
    5030-1

Spécifications

À propos de cet article

Designed for use with 96-Well thermal cyclers, Thin-Wall Micro Reaction Tubes and Strip Caps prevent cross-contamination and provide a secure seal for oil-free thermal cycling

Recommended maximum volume for thermal cycling amplifications is 100 µl. Maximum fill volume for post-thermal cycling reagent additions to permit mixing is 200 µl. Tubes and strip caps are supplied nonsterile, and are autoclavable at 121 °C (250 °F), 15psi, for fifteen minutes. Both strip tubes and strip caps may be easily cut. Individual 0.2 ml tubes require an 8 or 12 strip cap to seal each tube individually.

Tubes provide optimal contact with the sample block, thus ensuring optimal heat transfer. Tubes are fabricated from virgin polypropylene and feature a highly polished inner surface to allow for maximum sample recovery.

Ordering information: Cap strips should be ordered separately.
Frequently Bought Together