VWR® PCR Strip Tubes & Caps 8-Strip
# de catalogue 20170-004
Fournisseur: VWR International
Spécifications
- Cap Type:Domed
- Capacity:0.2 ml
- Format:Strip
- Color:Natural
- Description:Tube strips with separate bubble cap strips
- Environmentally Preferable:Sustainable Packaging,Low Manufacturing Impact
- Cat. no.:20170-004
Spécifications
A propos de cet article
Ideal for all PCR protocols, these 0.1 and 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
- Tubes and caps are chemically inert and autoclavable
- Smooth, thin, uniform walls for accurate thermal transfer
- Caps fit precisely to ensure a positive, contamination-free environment
Tube strips are certified free of RNase, DNase, and human DNA (lot-by-lot certificates available).
Ordering information: Tube strips with separate bubble cap strips are packed with 125 strips of 8 tubes and 125 strips of 8 caps in separate bags inside a box. Tube strips for real time PCR come with individually attached, optically clear caps.
Customers Who Bought This Also Bought