Nalgene® Utility Carrier, Thermo Scientific
# de catalogue 19757-060
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:HDPE utility carrier
- Cat. no.:19757-060
- L×W×H:37.5×24.1×15.3 cm (14¾×9½×6")
- Supplier No.:7130-0010
Spécifications
A propos de cet article
Carry glassware, samples, or tools with Nalgene® Utility Carriers
Carriers feature resilient construction with molded-in center divider and handle. Available in polyethylene or autoclavable polypropylene.
Customers Who Bought This Also Bought