ContactUsLinkComponent
VWR* Stackable Sharps Container Safety System
VWR® Sharps Container Systems, Multiple Colors and Styles
# de catalogue 19001-013
Fournisseur:  VWR International
Product available on GSA Advantage®
VWR* Stackable Sharps Container Safety System
VWR® Sharps Container Systems, Multiple Colors and Styles
# de catalogue 19001-013
Fournisseur:  VWR International
Product available on GSA Advantage®
Spécifications
  • Capacity:
    16 gal.
  • Dimensions:
    28.6W×28.6D×64.8H cm (11.25W×11.25D×25.5"H)
  • Color:
    Red
  • Cat. no.:
    19001-013
  • Style:
    Stackable
Spécifications
A propos de cet article
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps.
  • Puncture-resistant
  • Autoclave-safe and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports
The stackable sharps containers are puncture-resistant and both autoclaves-safe and incinerator-safe. The tortuous path systems restrict hand entry. The extended neck sharps containers feature a funneled vertical drop and extended handles.
Customers Who Bought This Also Bought