Laboratory Gas Lighters, G.C. Fuller
# de catalogue 18385-223
Fournisseur: G C Fuller
Spécifications
- Description:Replacement flint tips
- Cat. no.:18385-223
- Supplier No.:3001X
Spécifications
A propos de cet article
For lighting gas burners.
Designed with a rust-resistant zinc-plated spring handle and deep gas trap for quick ignitions. File rotates for fresh sparking surface. Each flint tip produces up to 2000 ignitions.
Customers Who Bought This Also Bought