Spécifications
- Packaging:Spray Bottle
- Description:RNase AWAY®
- Size:475 mL
- Cat. no.:17810-491
- Supplier No.:7002
Spécifications
A propos de cet article
RNase AWAY® eliminates RNase and DNA from laboratory surfaces
DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
RNase AWAY® eliminates RNase and DNA from laboratory surfaces
DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
Note: Should not be used as a reagent.
The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware, and plastic ware. The decontaminant leaves no residue to interfere with gel polymerization or staining and contains no strong acids