ContactUsLinkComponent
RNase AWAY*
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
# de catalogue 17810-491
Fournisseur:  Thermo Scientific
Numéro CAS:  
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
# de catalogue 17810-491
Fournisseur:  Thermo Scientific
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    Spray Bottle
  • Description:
    RNase AWAY®
  • Size:
    475 mL
  • Cat. no.:
    17810-491
  • Supplier No.:
    7002
Spécifications
A propos de cet article
RNase AWAY® eliminates RNase and DNA from laboratory surfaces
DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
RNase AWAY® eliminates RNase and DNA from laboratory surfaces
DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
Note: Should not be used as a reagent.
The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware, and plastic ware. The decontaminant leaves no residue to interfere with gel polymerization or staining and contains no strong acids
Customers Who Bought This Also Bought