Order Entry
ContactUsLinkComponent
BRUSH FOR PIPET
Pipette Cleaning Brush, Justman Brush
# de catalogue 17080-012
Fournisseur:  Justman Brush
Pipette Cleaning Brush, Justman Brush
# de catalogue 17080-012
Fournisseur:  Justman Brush

Spécifications

  • Bristle Material:
    Nylon
  • Brush Head Diameter:
    4.7 - 23.8 mm (3/16 - 15/16")
  • Description:
    Pipet Cleaning Brush
  • Total length:
    17"
  • Cat. no.:
    17080-012
  • Supplier No.:
    1872
  • Brush Head Length:
    7 1/2"

Spécifications

A propos de cet article

Brush features white nylon bristles and a tapered end, and is ideal for cleaning pipets.

Customers Who Bought This Also Bought