Nalgene® Right-To-Know Safety Wash Bottles, Thermo Scientific
# de catalogue 16651-991
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:28 mm
- Capacity:500 ml (16.9 oz.)
- Description:Assortment
- Cat. no.:16651-991
- Cap Color:—
- Supplier No.:2425-0500
Spécifications
A propos de cet article
Wash bottles are silk screened with high-quality, chemical-resistant inks.
To prevent cross-contamination, each bottle has a color bar that corresponds to its colored closure for easy hazard and chemical identification. These leakproof bottles meet requirements of OSHA Hazard Communication Regulation 29 CFR 1910.1200(f)(4).
All bottles except sodium hypochlorite bottles are molded of translucent LDPE for chemical resistance and durability, and include a polypropylene closure. Sodium hypochlorite bottles are molded of white LDPE for light-protective barrier qualities between 280 and 450 nm. They include an HDPE closure.
Ordering information: Assortment 16651-991 includes bottles (500 ml) and closures for all chemicals listed except sodium hypochlorite.
Customers Who Bought This Also Bought