VWR® Wide Mouth Wash Bottles, LDPE
# de catalogue 16650-162
Fournisseur: VWR International
Spécifications
- Cap Size:38 mm
- Capacity:250 ml (8.5 oz.)
- Cat. no.:16650-162
- Cap Color:Red
Spécifications
A propos de cet article
These bottles save time, help avoid mistakes, and increase safety.
- Color-coded caps provide quick, safe identification
- Wide-mouthed design
- One-piece cap and nozzle plus delivery tube
- Single drop or needle-thin stream
Color-coded, wide mouth bottles are ideal for the quick identification of the many special solutions and reagents needed in your lab. Bottles are durable, chemical-resistant, squeezable, and easy to clean. One-piece colored cap and nozzle are polypropylene. Delivery tube is polyethylene. The leakproof wash cap assembly will deliver a single drop or a needle thin stream, with finger-tip squeeze control of flow. For a coarser flow, just cut the nozzle tip back.
Customers Who Bought This Also Bought