ContactUsLinkComponent
KIMAX* Specific Gravity Bottle
KIMAX® Specific Gravity Bottle, Hubbard, Kimble Chase
# de catalogue 16620-004
KIMAX* Specific Gravity Bottle
KIMAX® Specific Gravity Bottle, Hubbard, Kimble Chase
# de catalogue 16620-004
Spécifications
  • Capacity:
    24 ml (0.81 oz.)
  • ST Stopper No.:
    24/12
  • Supplier No.:
    15110-24
Spécifications
A propos de cet article
For specific gravity of road oils, road tars, soft tar pitches, and asphalt cements according to ASTM D70, AASHTO T43; drying oils, varnishes, resins, and related materials (ASTM D1963); water and brine (ASTM D1429); and insulating varnishes (ASTM D115)
Stopper is concave on underside and has hole, approx. 1.6 mm in diameter, to allow air to escape.
Customers Who Bought This Also Bought