Snap Caps, Polyethylene, Kimble Chase
# de catalogue 16358-270
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Size:5
- Description:Snap Caps
- Cat. no.:16358-270
- For Use With:200 ml, 250 ml, 500 ml Flasks
- Supplier No.:28150R-5
Spécifications
A propos de cet article
Replacement snap caps for the 89001-852 and -870 series volumetric flasks.
Customers Who Bought This Also Bought